Please login to your online account to display your discounted pricing

Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts


Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Manufacturer: thermo scientific™  62251*

 View more versions of this product

Catalog No. PI62261

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) myECL Imager
Formulation Black stylus resembling a pen; and a round plastic clip to mount on the side of the imager
Quantity 1 set
Item Description Touchscreen Stylus and Holder
Description Touchscreen Stylus and Holder
Sufficient For Replacement of myECL Imager accessory; for interaction with the imager touchscreen

Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.