Please login to your online account to display your discounted pricing

Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts


Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Manufacturer: thermo scientific™  62243

 View more versions of this product

Catalog No. PI62243

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) myECL™ Imager
For Use With (Application) myECL Imager
Description White Light Conversion Screen
Formulation White glass sample screen to cover the transilluminator; 24.5cm x 19.5cm x 3.2mm
Quantity 1 screen
Sufficient For Replacement of myECL Imager accessory; used on the transilluminator for acquiring images in the epi-white (visible mode)
Item Description White Light Conversion Screen

Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.