Please login to your online account to display your discounted pricing

Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts


Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Manufacturer: thermo scientific™  62241

 View more versions of this product

Catalog No. PI62241

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) myECL™ Imager
For Use With (Application) myECL Imager
Item Description Blue Filter
Sufficient For Addition of a blue filter to one of two open positions in the myECL Imager for detection of blue-fluorescent dyes and stains
Formulation 445-480nm bandpass filter; threaded, 50mm-diameter glass filter
Description Blue Filter
Quantity 1 filter

Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.