Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts
Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.
Manufacturer:  Thermo Scientific™ 62240
 View more versions of this product
Catalog No. PI62240
Specifications
Description
Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.
Replacement parts for the myECL Imager and accessories to expand upon its capabilities.
Description & Specifications
Specifications
Red Filter
600-640nm bandpass filter; threaded, 50mm-diameter glass filter
Addition of a red filter to one of two open positions in the myECL Imager for detection (emission) wavelengths of popular red or purple fluorescent protein stains
myECL Imager
1 filter
Red Filter