Please login to your online account to display your discounted pricing

Thermo Scientific™ myECL™ Imager Accessories and Replacement Parts


Document and analyze images of blots and gels using a sensitive and easy-to-use system with an intuitive touchscreen interface and advanced integrated software.

Manufacturer: thermo scientific™  62238

 View more versions of this product

Catalog No. PI62238

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) myECL Imager
Description Orange Filter
Sufficient For Replacement for the pre-installed orange filter in the myECL Imager for detection of colorimetric protein stains (silver, coomassie) and wavelengths of common fluorescent nucleic acid gel stains (e.g., ethidium bromide) and protein stains
Formulation 565-620nm bandpass filter; threaded, 50mm-diameter glass filter
Quantity 1 filter
Item Description Orange Filter

Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.

Replacement parts for the myECL Imager and accessories to expand upon its capabilities.