Invitrogen™ MUSK Recombinant Human Protein DFS Item
MUSK plays an important and selective role in many aspects of skeletal muscle development and function.
Manufacturer:  Invitrogen™ LSPV4113
 View more versions of this product
Catalog No. PV4113
Description
Description
MUSK plays an important role in skeletal muscle development through the initial step of postsynaptic differentiation and formation of end-plate bands in cooperation with Rapsyn.
Specifications
Specifications
NP_005583.1
Frozen
69.4kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
Kinase Assay
MUSK Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
1mg
RUO - research use only
MUSK
Baculovirus
Human
GST Tag
Documents