Please login to your online account to display your discounted pricing

Alfa Aesar™ Muscone, 99%


Quantity 100mg

Manufacturer: alfa aesar™  J67481MC

 View more versions of this product

Catalog No. AAJ67481MC

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 238.41
Chemical Name or Material Muscone
Physical Form Oil
Assay 99%
MDL Number MFCD00211114
TSCA TSCA
Synonym 3-Methylcyclopentadecanone
Quantity 100mg
RTECS Number GY0950000
CAS 541-91-3
Color Colorless

Specifications

Chemical Name or Material Muscone
CAS 541-91-3
Formula Weight 238.41
MDL Number MFCD00211114
Synonym 3-Methylcyclopentadecanone