Please login to your online account to display your discounted pricing

Alfa Aesar™ Muscone, 99%

Chemical Identifiers

Chemical Name or Material Muscone
CAS 541-91-3
Formula Weight 238.41
MDL Number MFCD00211114
Synonym 3-Methylcyclopentadecanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67481MC

 
alfa aesar™
J67481MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67481ME

 
alfa aesar™
J67481ME
500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 238.41
Chemical Name or Material Muscone
Physical Form Oil
Assay 99%
MDL Number MFCD00211114
TSCA TSCA
Synonym 3-Methylcyclopentadecanone
Quantity 100mg
RTECS Number GY0950000
CAS 541-91-3
Color Colorless