Please login to your online account to display your discounted pricing

Alfa Aesar™ Muscimol, 99+%

Selective GABA-a agonist

Chemical Identifiers

Chemical Name or Material Muscimol
CAS 2763-96-4
Molecular Formula C4H6N2O2
Formula Weight 114.1
MDL Number MFCD00057894
UN Number UN1544
Beilstein 1618960
Merck Index 14,6312
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ61720MA

  SDS
alfa aesar™
J61720MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Muscimol
CAS 2763-96-4
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
RTECS Number NY3325000
TSCA TSCA
Physical Form Powder
Assay ≥99%
Molecular Formula C4H6N2O2
Formula Weight 114.1
MDL Number MFCD00057894
UN Number UN1544
Beilstein 1618960
Merck Index 14,6312
Color White to Off-white
Quantity 10mg

Applications
Muscimol has been widely employed for this agonist activity in studies probing the consequent effects of GABAA stimulation. It is also used as a potent and specific GABAA receptor agonist/GABA analog. It also inhibits memory retention and attenuates airway constriction in vivo.

Solubility
Soluble in water (10 mg/ml), methanol, ethanol, 0.05 M HCl (20 mg/ml), DMSO (sparingly soluble), DMF (sparingly soluble), and PBS (pH 7.2) (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents, strong acids, acid chlorides, acid anhydrides.