Please login to your online account to display your discounted pricing

Musashi 2 Mouse anti-Human, Mouse, eFluor 660, Clone: C1, eBioscience™

Mouse Monoclonal Antibody

$160.95 - $390.05

Specifications

Antigen Musashi 2
Clone C1
Host Species Mouse
Species Reactivity Human, Mouse
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124514

 
life technologies
LS50967780
25μg Each for $160.95

501124515

 
life technologies
LS50967782
100μg Each for $390.05
Description & Specifications

Specifications

Antigen Musashi 2
Clone C1
Host Species Mouse
Species Reactivity Human, Mouse
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The monoclonal antibody C1 recognizes human and mouse Musashi-2 (Msi2), a highly conserved RNA-binding protein. Musashi-2 contains two RNA-recognition domains and is expressed as one of two isoforms of 36 and 37kDa. Musashi 2 with its family member Musashi-1, are expressed at high levels in stem and tumor cells. During neural development, Musashi-2 is expressed in progenitor cells in the ventricular and subventricular zones, while postnatally, Musashi-2 expression is limited to multiple subsets of neurons and astrocytes in addition to adult stem cell populations. Functionally, Musashi-2 associates with Sox2 and is required for self-renewal of embryonic stem cells. Much like its role in maintaining stemness in neural progenitors, Musashi-2 is expressed in hematopoetic stem cells and plays a similar role in maintaining self-renewal. Musashi-2 has been found to be overexpressed in human myeloid leukemia and the levels of expression correlate with decreased survival. Musashi-2 may serve as a prognostic marker for acute myeloid leukemia (AML).