missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Musashi-2, eFluor 660, clone: C1, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS50967782

 View more versions of this product

Catalog No. 501124515



Description

Description

Msi2 (musashi homolog 2), also known as MSI2H, is a 328 amino acid protein that localizes to the cytoplasm and contains two RRM (RNA recognition motif) domains. Expressed ubiquitously at low levels, Msi2 functions as an RNA binding protein that, by regulating the expression of target mRNAs, is thought to play a role in the proliferation and maintenance of stem cells within the central nervous system. Msi2 is subject to posttranslational phosphorylation and is upregulated in response to brain injury, suggesting a role in healing and brain tissue regeneration. Chromosomal aberrations involving the Msi2 gene are associated with the progression of chronic myeloid leukemia. Multiple isoforms of Msi2 exist due to alternative splicing events.Tissue specificity: Ubiquitous; detected at low levels.

Description: The monoclonal antibody C1 recognizes human and mouse Musashi-2 (Msi2), a highly conserved RNA-binding protein. Musashi-2 contains two RNA-recognition domains and is expressed as one of two isoforms of 36 and 37kDa. Musashi 2 with its family member Musashi-1, are expressed at high levels in stem and tumor cells. During neural development, Musashi-2 is expressed in progenitor cells in the ventricular and subventricular zones, while postnatally, Musashi-2 expression is limited to multiple subsets of neurons and astrocytes in addition to adult stem cell populations. Functionally, Musashi-2 associates with Sox2 and is required for self-renewal of embryonic stem cells. Much like its role in maintaining stemness in neural progenitors, Musashi-2 is expressed in hematopoetic stem cells and plays a similar role in maintaining self-renewal. Musashi-2 has been found to be overexpressed in human myeloid leukemia and the levels of expression correlate with decreased survival. Musashi-2 may serve as a prognostic marker for acute myeloid leukemia (AML).Applications Reported: This C1 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.Applications Tested: This C1 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using high pH antigen retrieval and can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Musashi-2
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q920Q6, Q96DH6
MSI2H, RNA-binding protein Musashi homolog 2, musashi-2
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Murine
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
C1
eFluor 660
MSI2
Liquid
Msi2
IgG3, kappa, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
124540, 76626
Documents

For Research Use Only.