Please login to your online account to display your discounted pricing

Musashi-1 Rat, Clone: 14H1, eBioscience™

Rat Monoclonal Antibody

$143.55 - $346.55

Specifications

Antigen Musashi-1
Clone 14H1
Host Species Rat
Species Reactivity Human, Mouse, Rat
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013834

 
life technologies
LS14989680
25μg Each for $143.55

5013835

 
life technologies
LS14989682
100μg Each for $346.55
Description & Specifications

Specifications

Antigen Musashi-1
Clone 14H1
Host Species Rat
Species Reactivity Human, Mouse, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C. Avoid repeated freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This 14H1 monoclonal antibody recognizes human Musashi-1 (MSI-1), a 39kDa RNA binding protein. Musashi-1 is expressed in CNS stem and progenitor cells with decreased expression in differentiating cells. Musashi-1 functions as a translational repressor, directly regulating its target proteins, Numb and p21 (CIP1). Musashi-1 has been shown to be expressed in stem cells in a variety of other tissues such as the gut, stomach, mammary gland, and hair follicles. In addition to normal tissue expression, Musashi-1 expression has been identified in cells from several tumor types, mainly pulmonary and adenocarcinoma, as well as large and small cell carcinomas. Recently, expression of Musashi-1 in ischemic lesions following stroke provides evidence for neurogenesis as well as the role of Musashi-1 in regulating apoptosis.

Musashi-1 is highly conserved and the 14H1 antibody also recognizes mouse and rat Musashi-1.