missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Murexide, TCI America™

Manufacturer:  TCI America M04745G

 View more versions of this product

Catalog No. M04745G



Specifications

Specifications

Murexide
C8H3N5O6-2
Ammonium Purpurate
C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O
265.141
284.19
Purple-Red
3051-09-0
MFCD00012777
MUMMBXDKKJEASE-UHFFFAOYSA-L
6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
51381221
Crystal-Powder at 20°C
5g

Chemical Identifiers

3051-09-0
265.141
MUMMBXDKKJEASE-UHFFFAOYSA-L
51381221
C1(=C(N=C(NC1=O)[O-])[O-])N=C2C(=O)NC(=O)NC2=O
C8H3N5O6-2
MFCD00012777
Ammonium Purpurate
6-oxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1H-pyrimidine-2,4-diolate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents