missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Murexide (Indicator Tablets), Fisher Chemical

Manufacturer:  Fisher Scientific Company M2731000

 View more versions of this product

Catalog No. M273-1000



 

Specifications Description & Specifications

Specifications

Murexide
Pass Test
Solid
C8H8N6O6
284.19
770°C
Ammonium Purpurate Acid
3051-09-0,7447-40-7
Glass Bottle
1.9g/cm³
Off-white
MFCD00012777
1000 tablets

Chemical Identifiers

3051-09-0,7447-40-7
C8H8N6O6
MFCD00012777
Murexide
Solid
284.19
Ammonium Purpurate Acid

Safety and Handling

SDS & Safety Documents