Please login to your online account to display your discounted pricing

Murexide (Indicator Tablets), Fisher Chemical


Quantity 100 tablets
Packaging Glass Bottle

Manufacturer: fisher scientific company  M273100

 View more versions of this product

Catalog No. M273-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Murexide
CAS 3051-09-0
ChemAlert Storage Symbol Gray
CAS 7447-40-7
Identification Pass Test
Packaging Glass Bottle
Physical Form Solid
Density 1.9g/cm³
Molecular Formula C8H8N6O6
Color Off-white
Formula Weight 284.19
MDL Number MFCD00012777
Melting Point 770°C
Quantity 100 tablets
Synonym Ammonium Purpurate Acid

Specifications

Chemical Name or Material Murexide
CAS 3051-09-0
CAS 7447-40-7
Physical Form Solid
Molecular Formula C8H8N6O6
Formula Weight 284.19
MDL Number MFCD00012777
Synonym Ammonium Purpurate Acid
Accessories
 

SDS & Safety Documents