Please login to your online account to display your discounted pricing

Murexide, ACS Grade, LabChem™

$133.00 - $133.00

Chemical Identifiers

Chemical Name or Material Murexide
CAS 3051-09-0
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C8H8N6O6
Formula Weight 284.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC174907

 
lab chem
LC174907
Amber Glass 5g Each for $133.00
Description & Specifications

Specifications

Formula Weight 284.18
CAS 3051-09-0
Health Hazard 2 GHS H Statement
Product is not hazardous.
Quantity 5g
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 100
Purity Grade ACS Grade
Physical Form Powder
Packaging Amber Glass
Melting Point 300°C
Color Maroon
Identification Passes Test
Recommended Storage Room Temperature
Solubility Soluble in water
Linear Formula C8H8N6O6
Chemical Name or Material Murexide
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature