missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Murexide, ACROS Organics™

$53.45 - $171.88

Chemical Identifiers

CAS 3051-09-0
Molecular Formula C8H8N6O6
Formula Weight 284.19
MDL Number MFCD00012777
Chemical Name or Material Murexide
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC206890050 SDS Fisher Scientific Acros
AC206890050
Glass bottle 5g Each for $53.45
AC206890250 SDS Fisher Scientific Acros
206890250
Glass bottle 25g Each for $171.88
AC206891000 SDS Thermo Fisher Scientific
AC206891000
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC206895000 SDS Thermo Fisher Scientific
AC206895000
Glass bottle 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Indicator
Description & Specifications

Chemical Identifiers

3051-09-0
284.19
Murexide
C8H8N6O6
MFCD00012777
ACS Reagent Grade

Specifications

3051-09-0
Glass bottle
Crystals and/or Chunks
Colorless to white
MFCD00012777
Murexide
Authentic
Suitability as a complexometric indicator: passes
C8H8N6O6
284.19
5g
ACS Reagent Grade

Safety and Handling

EINECSNumber : 221-266-6

TSCA : TSCA