Please login to your online account to display your discounted pricing

Murexide, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  206890250

 View more versions of this product

Catalog No. AC206890250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals and/or Chunks
Purity Grade ACS Reagent Grade
Formula Weight 284.19
Quantity 25g
Color Colorless to white
TSCA TSCA
EINECS Number 221-266-6
Packaging Glass bottle
Infrared Spectrum Authentic
Suitability Suitability as a complexometric indicator: passes
MDL Number MFCD00012777
CAS 3051-09-0
Chemical Name or Material Murexide
Molecular Formula C8H8N6O6

Specifications

Chemical Name or Material Murexide
CAS 3051-09-0
Purity Grade ACS Reagent Grade
Molecular Formula C8H8N6O6
Formula Weight 284.19
MDL Number MFCD00012777