Thermo Scientific™ Multisip™ Split Septum Plugs
Thermo Scientific™ MULTISIP™ Split Septum Plugs feature a unique proprietary split septum design
$1,691.94 - $3,081.06
Specifications
Material Inert polymer
Pierceable Yes
Sterile No
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
AB-1174 Thermo Scientific™
AB1174
10 Trays of 96 plugs Pack of 960 Each for $3,081.06
AB-0972 Thermo Fisher Scientific
AB0972
Case of 1000 Plugs Case of 1000 Each for $1,691.94
Specifications
Description
Thermo Scientific™ MULTISIP™ Split Septum Plugs feature a unique proprietary split septum design.
  • Individual plugs made from an inert polymer
  • Pierceable with standard pipetting equipment
  • 96 plugs can be pierced simultaneously
  • 100% resealing even after 200 piercings
  • Less than 4N pierce pressure
  • Solvent resistant, including acetonitrile and DMSO (for long-term storage with DMSO, samples should be stored at 4°C or below)
  • Temperature range -170°C to +120°C; fully autoclavable
  • Apply 96 plugs individually with the Cap-It tool
Description & Specifications
Specifications
Inert polymer
Yes
No