Multisip™ Split Septum Plugs
Thermo Scientific™ MULTISIP™ Split Septum Plugs feature a unique proprietary split septum design
Manufacturer:  Thermo Fisher Scientific AB0972
 View more versions of this product
Catalog No. AB-0972
Specifications
Description
Thermo Scientific™ MULTISIP™ Split Septum Plugs feature a unique proprietary split septum design.
  • Individual plugs made from an inert polymer
  • Pierceable with standard pipetting equipment
  • 96 plugs can be pierced simultaneously
  • 100% resealing even after 200 piercings
  • Less than 4N pierce pressure
  • Solvent resistant, including acetonitrile and DMSO (for long-term storage with DMSO, samples should be stored at 4°C or below)
  • Temperature range -170°C to +120°C; fully autoclavable
  • Apply 96 plugs individually with the Cap-It tool
Description & Specifications
Specifications
Inert polymer
No
7mm; Case of 1000 Plugs
1000
Yes
Case of 1000 Plugs
1000