Arcor Electronics Multiple-Use Bare Copper Wire
Solid soft drawn annealed copper wire
Manufacturer:  Arcor Electronics BARE16GA
 View more versions of this product
Catalog No. 15-545-1A
Description
Description
  • Available in various tempers and sizes
  • Uninsulated
For busbar crafts, metal finishers' hanging wire, and grounding applications.
Compliance
Meets QQ-W-343, TYPE S, MIL-W-3861, TYPE S, ASTM™-B-3.
Specifications
Specifications
QQ-W-343, TYPE S, MIL-W-3861, TYPE S, ASTM-B-3
1.3mm
16
0.25 lb.
Copper
0.05 in.
For busbar crafts, Metal finishers hanging wire and grounding applications
126 Ft./lb.
113g
Bare Copper Wire
Documents