Arcor Electronics Multiple-Use Bare Copper Wire DFS Item
Solid soft drawn annealed copper wire
Manufacturer:  Arcor Electronics AWC34164
 View more versions of this product
Catalog No. 15-545-2A
Description
Description
Solid soft drawn annealed copper wire
  • Available in various tempers and sizes
  • Uninsulated
For busbar crafts, metal finishers' hanging wire, and grounding applications.
Compliance
Meets QQ-W-343, TYPE S, MIL-W-3861, TYPE S, ASTM™-B-3.
Specifications
Specifications
Bare Copper Wire
454 g
126 ft./lb.
For busbar crafts, Metal finishers hanging wire and grounding applications
0.05 in.
Copper
1 lb.
16
1.3 mm
QQ-W-343, TYPE S, MIL-W-3861, TYPE S, ASTM-B-3