Please login to your online account to display your discounted pricing

Alfa Aesar™ MultiMod PTM Mass Spec Certified Standard I

For cellular and molecular biology applications

Specifications

Product Type MultiMod PTM Mass Spec Certified Standard I, 500pmol each peptide per vial 1 vial
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Phosphorylated Angiotensin II, Glycosylated Erythropoietin (117-131), Sulfated Leu-Enkephalin, Nitrosylated eNOS (205-21
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67741LPL

 
alfa aesar™
J67741LPL
1 vial, 500 pmol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67741ODL

 
alfa aesar™
J67741ODL
3 vials, 500 pmol/vial CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type MultiMod PTM Mass Spec Certified Standard I, 500pmol each peptide per vial 1 vial
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence Phosphorylated Angiotensin II, Glycosylated Erythropoietin (117-131), Sulfated Leu-Enkephalin, Nitrosylated eNOS (205-21
Regulatory Status RUO
Purification Method HPLC