Please login to your online account to display your discounted pricing

Alfa Aesar™ MultiMod PTM Mass Spec Certified Standard I


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67741LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67741LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements -30° to -10°C
Purification Method HPLC
Regulatory Status RUO
Sequence Phosphorylated Angiotensin II, Glycosylated Erythropoietin (117-131), Sulfated Leu-Enkephalin, Nitrosylated eNOS (205-21
Product Type MultiMod PTM Mass Spec Certified Standard I, 500pmol each peptide per vial 1 vial
Format Lyophilized powder
Quantity 1 vial, 500 pmol