Please login to your online account to display your discounted pricing

Thermo Scientific Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

$1,843.48 - $1,843.48

Specifications

Autoclavable 50 times at 121°C, 1 bar, 20 min.
Compatibility Multidrop Combi, Multidrop 384, Multidrop DW
Diameter Tip: 0.5mm; Tubing: 1.3mm
Dispensing Range 5 to 2500µL
Material Tip: PP; Tubing: Silicone
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Description Price Quantity    

50-950-366

 
thermo scientific
24070290
Tubing set for standard tube dispensing cassette (with 40 cm tubing set Each for $1,843.48
Description & Specifications

Specifications

Autoclavable 50 times at 121°C, 1 bar, 20 min.
Compatibility Multidrop Combi, Multidrop 384, Multidrop DW
Diameter Tip: 0.5mm; Tubing: 1.3mm
Dispensing Range 5 to 2500µL
Material Tip: PP; Tubing: Silicone

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

Ordering Information:

Cat Nos 24070297 and 24070290 include tip combs.