Please login to your online account to display your discounted pricing

Alfa Aesar™ Multi Ion anion IC standard solution, Specpure™ (Waters) in H2O

Chemical Identifiers

Chemical Name or Material Multi Ion anion IC standard solution
Name Note Specpure™ (Waters) in H2O
Purity Grade Specpure™
MDL Number MFCD00144814
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35564AE

 
alfa aesar™
35564AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Multi Ion anion IC standard solution
Quantity 100mL
TSCA TSCA
Name Note Specpure™ (Waters) in H2O
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00144814