Please login to your online account to display your discounted pricing

MULT1 (NKG2D Ligand) Armenian Hamster anti-Mouse, Rat, Clone: 5D10, eBioscience™

Armenian Hamster Monoclonal Antibody

$114.55 - $187.05

Specifications

Antigen MULT1 (NKG2D Ligand)
Clone 5D10
Host Species Armenian Hamster
Gene Alias NKG2DL, MULT-1
Species Reactivity Mouse, Rat
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5013228

 
life technologies
LS14586381
50μg Each for $114.55

5013229

 
life technologies
LS14586382
100μg Each for $187.05
Description & Specifications

Specifications

Antigen MULT1 (NKG2D Ligand)
Clone 5D10
Host Species Armenian Hamster
Gene Alias NKG2DL, MULT-1
Species Reactivity Mouse, Rat
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 5D10 monoclonal antibody reacts with the murine MULT-1 (Murine ULBP-Like Transcript 1). MULT-1 is a type I transmembrane protein with two Ig-like domain and is one of the known murine NKG2-D ligands that include RAE1 molecules (alpha, beta, epsilon, gamma and delta) and H60. MULT-1 is a high affinity ligand for NKG2D.Expression of NKG2-D ligands is low or absent on normal adult tissues. However, stressed or transformed cells express NKG2-D ligand and can activate NK cell tumoricidal activity. Until now, the expression of NKG2-D ligands has been mainly studied with NKG2-D tetramers recognizing all NKG2-D ligands.