Please login to your online account to display your discounted pricing

Mud Puppies

Necturus maculosus.

$153.90 - $172.80

Specifications

Length (English) 10 to 14 in.
Preservative Bio-Fresh™
Product Type Necturus
Quantity 150/CS
Length (Metric) 25.4 to 35.5cm
View More Specs
Products
Catalog Number Mfr. No. Injection Type Price Quantity    

S5111S

 
fisher science education
136102210
Double Pack of 10 Each for $153.90
Case of 15 Pack for $2,308.50

S5112S

 
fisher science education
136103010
Triple Pack of 10 Each for $172.80
Case of 15 Pack for $2,592.00
Description & Specifications

Specifications

Length (English) 10 to 14 in.
Preservative Bio-Fresh™
Product Type Necturus
Quantity 150/CS
Length (Metric) 25.4 to 35.5cm
Size Large

Necturus, a freshwater salamander, is commonly known by its nonscientific name of “mud puppy.”

  • Unusual, unique gills
  • Distinctively long tail, used for swimming
  • Large specimens (10 to 14 in.) supplied in Fisher Science Education™ Bio-Fresh™

Sold in packs of 10.