Learn More
Mucochloric acid, 99%, Thermo Scientific Chemicals
$188.80 - $188.80
Chemical Identifiers
CAS | 87-56-9 |
---|---|
Molecular Formula | C4H2Cl2O3 |
Molecular Weight (g/mol) | 168.96 |
MDL Number | MFCD00006966,MFCD00006966 |
InChI Key | LUMLZKVIXLWTCI-IHWYPQMZSA-N |
Synonym | mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597 |
PubChem CID | 2771871 |
IUPAC Name | (Z)-2,3-dichloro-4-oxobut-2-enoic acid |
SMILES | OC(=O)C(\Cl)=C(\Cl)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC128065000
|
Thermo Scientific Chemicals
128065000 |
500 g |
Each for $188.80
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87-56-9 | |
168.96 | |
LUMLZKVIXLWTCI-IHWYPQMZSA-N | |
2771871 | |
OC(=O)C(\Cl)=C(\Cl)C=O |
C4H2Cl2O3 | |
MFCD00006966,MFCD00006966 | |
mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597 | |
(Z)-2,3-dichloro-4-oxobut-2-enoic acid |
Specifications
87-56-9 | |
100.0 | |
White to Beige | |
Authentic | |
Plastic bottle | |
ClC(CHO)=C(Cl)CO2H | |
500 g | |
15, 6383 | |
Solubility in water: soluble in hot water. Other solubilities: slightly soluble in cold water, soluble in hot benzene and alcohol | |
OC(=O)C(\Cl)=C(\Cl)C=O | |
168.96 | |
168.96 | |
Crystalline Powder |
98.5 | |
125.0°C to 128.0°C | |
186°C | |
99% | |
C4H2Cl2O3 | |
MFCD00006966,MFCD00006966 | |
03, 727 | |
mucochloric acid, 2,3-dichloromaleic aldehyde acid, kyselina mukochlorova, 2z-2,3-dichloro-4-oxobut-2-enoic acid, 2-butenoic acid, 2,3-dichloro-4-oxo-, 2z, aldehydodichloromaleic acid, dichloromalealdehydic acid, unii-5i5877jhiw, kyselina mukochlorova czech, ccris 6597 | |
LUMLZKVIXLWTCI-IHWYPQMZSA-N | |
(Z)-2,3-dichloro-4-oxobut-2-enoic acid | |
2771871 | |
99% | |
Mucochloric acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Toxic if swallowed.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
GHS Signal Word: Danger
EINECSNumber : 201-752-4
RUO – Research Use Only