missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mucochloric acid, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 128065000

 View more versions of this product

Catalog No. AC128065000


 

Specifications Description & Specifications

Specifications

≥98.5% (on dry substance)
168.96
White to light beige
03, 727
100.0
Plastic bottle
Authentic
99%
C4H2Cl2O3
MFCD00006966
Crystalline Powder
500g
15, 6383
125° to 128°C
4% max. (K.F.)
ClC(CHO)=C(Cl)CO2H
Mucochloric acid
87-56-9
98.5
2, 3-Dichloro-4-oxo-2-butenoic acid

Chemical Identifiers

87-56-9
100.0
C4H2Cl2O3
168.96
2, 3-Dichloro-4-oxo-2-butenoic acid
≥98.5% (on dry substance)
98.5
99%
ClC(CHO)=C(Cl)CO2H
MFCD00006966
Mucochloric acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Toxic if swallowed.
May cause respiratory irritation.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Obtain special instructions before use.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger