missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mucobromic acid, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC128055000

 View more versions of this product

Catalog No. AC128055000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥99.0%
257.86
White to beige
03, IV, 1721
100.0
Glass bottle
Mucobromic acid
99+%
Authentic
Crystalline Powder
500g
122° to 125°C
99.0
BrC(CHO)=C(Br)CO2H
C4H2Br2O3
488-11-9
MFCD00063745

Chemical Identifiers

488-11-9
100.0
C4H2Br2O3
257.86
Mucobromic acid
99.0
99+%
BrC(CHO)=C(Br)CO2H
MFCD00063745
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger