Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody.

$236.61 - $1,097.47

Specifications

Antigen Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1
Clone 45M1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

MS145P

 
thermo scientific™ lab vision™
MS145P
Purified Ab with BSA and Azide 1mL Each for $983.33

MS145P0

 
thermo scientific™ lab vision™
MS145P0
Purified Ab with BSA and Azide 100µL Each for $236.61

MS145P1

 
thermo scientific™ lab vision™
MS145P1
Purified Ab with BSA and Azide 500µL Each for $652.00

MS145P1ABX

 
thermo scientific™ lab vision™
MS145P1ABX
Ab without BSA/Azide (Frozen) 100µL Each for $770.22

MS145PABX

 
thermo scientific™ lab vision™
MS145PABX
Ab without BSA/Azide (Frozen) 200µL Each for $1,097.47

MS145R7

 
thermo scientific™ lab vision™
MS145R7
Ready-to-use for IHC (UVLP) 7mL Each for $571.18
Description & Specifications

Specifications

Antigen Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1
Clone 45M1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Host Species Mouse
Regulatory Status IVD
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 5AC (MUC5AC)/Gastric Mucin Ab-1, Mouse Monoclonal Antibody.

Mucins are high molecular eight glycoproteins with 80% carbohydrate contents and the remaining 20% is constituted by protein core. Gastric mucin is found in normal human gastrointestinal tract. Resurgence of gastric mucin reactivity during colonic carcinogenesis is due to re-expression of the peptide core of gastric (or fetal colonic) mucins.

Host Species: Mouse

Clone: 45M1

Isotype: IgG1/κ

Species Reactivity: Human, Monkey, Rabbit, Cat, Mouse, Rat, Pig, Hedgehog and Chicken. Does not react with cow. Others not known.

Epitope: Destroyed by β-mercaptoethanol and proteases but not by periodate treatment

Immunogen: M1 mucin preparation from the fluid of an ovarian mucinous cyst belonging to an O Le(a-b-) patient

Molecular Weight: >1000kDa

Positive Control: Stomach

Cellular Localization: Cytoplasmic and cell surface

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)