Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody.

$306.31 - $1,505.55

Specifications

Antigen Mucin 3 (MUC3) Ab-2
Clone 1143/B7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1734P

 
thermo scientific™ lab vision™
MS1734P
200μg/mL 1mL Each for $1,424.16

MS1734P0

 
thermo scientific™ lab vision™
MS1734P0
200μg/mL 100µL Each for $306.31

MS1734P1

 
thermo scientific™ lab vision™
MS1734P1
200μg/mL 500µL Each for $861.92

MS1734P1ABX

 
thermo scientific™ lab vision™
MS1734P1ABX
1mg/mL 100µL Each for $1,001.76

MS1734PABX

 
thermo scientific™ lab vision™
MS1734PABX
1mg/mL 200µL Each for $1,505.55
Description & Specifications

Specifications

Antigen Mucin 3 (MUC3) Ab-2
Clone 1143/B7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen MUC3 tandem repeat peptide
Isotype IgG2a

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 3 (MUC3) Ab-2, Mouse Monoclonal Antibody.

Mucin 3 (MUC3) is a glycoprotein. Distributed in colon and rectum, and to a lesser extent in breast, lung and salivary gland tissues.

Host Species: Mouse

Clone: 1143/B7

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Tandem repeats of the human MUC3

Immunogen: MUC3 tandem repeat peptide

Molecular Weight: High

Positive Control: Colon CA

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO