missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mucin 2 (MUC2) Ab-3, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 2 (MUC2) Ab-3, Mouse Monoclonal Antibody.

$309.31 - $1,505.55

Specifications

Antigen Mucin 2 (MUC2) Ab-3
Clone 996/1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS1729P Thermo Scientific™ Lab Vision™
MS1729P
200μg/mL 1mL Each for $1,424.16
MS1729P0 Thermo Scientific™ Lab Vision™
MS1729P0
200μg/mL 100μL Each for $309.31
MS1729P1 Thermo Scientific™ Lab Vision™
MS1729P1
200μg/mL 500μL Each for $871.20
MS1729P1ABX Thermo Scientific™ Lab Vision™
MS1729P1ABX
1mg/mL 100μL Each for $991.16
MS1729PABX Thermo Scientific™ Lab Vision™
MS1729PABX
1mg/mL 200μL Each for $1,505.55
Specifications

Description

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 2 (MUC2) Ab-3, Mouse Monoclonal Antibody.

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.

Host Species: Mouse

Clone: 996/1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Tandem repeats of the human MUC2

Immunogen: MUC2 tandem repeat peptide

Molecular Weight: 520kDa

Positive Control: Small intestine

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

Mucin 2 (MUC2) Ab-3
Immunohistochemistry (Paraffin)
Unlabeled
Murine
Cancer and Tumor Biology
MUC2 tandem repeat peptide
996/1
Monoclonal
Concentrated
RUO
Primary
IgG1