Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Mucin 2 (MUC2) Ab-3, Mouse Monoclonal Antibody


Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 2 (MUC2) Ab-3, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS1729P1ABX

 View more versions of this product

Catalog No. MS1729P1ABX

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry (Paraffin)
Isotype IgG1
Quantity 100µL
Regulatory Status RUO
Clone 996/1
Cross Reactivity Human
Antigen Mucin 2 (MUC2) Ab-3
Format Concentrated
Host Species Mouse
Immunogen MUC2 tandem repeat peptide
Research Discipline Cancer and Tumor Biology
Type Primary
Concentration 1mg/mL
Conjugate Unlabeled
Classification Monoclonal

Deliver accurate, reproducible results with the Thermo Scientific™ Mucin 2 (MUC2) Ab-3, Mouse Monoclonal Antibody.

Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.

Host Species: Mouse

Clone: 996/1

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Tandem repeats of the human MUC2

Immunogen: MUC2 tandem repeat peptide

Molecular Weight: 520kDa

Positive Control: Small intestine

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry

Note:

USA: RUO; Int'l: RUO