Please login to your online account to display your discounted pricing

Alfa Aesar™ MUC5AC 3, Certified Mass Spec grade

For research use

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements -30° to -10°C
Product Type MUC5AC 3, Certified Mass Spec grade
Format Lyophilized powder
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67801LPL

 
alfa aesar™
J67801LPL
1 vial CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67801ODL

 
alfa aesar™
J67801ODL
3 vials CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formulation Lyophilized powder
Recombinant No
Storage Requirements -30° to -10°C
Product Type MUC5AC 3, Certified Mass Spec grade
Format Lyophilized powder
Regulatory Status RUO