Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MUC-1, Mouse Monoclonal Antibody

Deliver accurate, reproducible results with the Thermo Scientific™ MUC-1, Mouse Monoclonal Antibody.

$264.02 - $1,278.86

Specifications

Antigen MUC-1
Clone SPM492
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MS10340P

 
thermo scientific™ lab vision™
MS10340P
1mL Each for $1,278.86

MS10340P0

 
thermo scientific™ lab vision™
MS10340P0
100µL Each for $264.02

MS10340P1

 
thermo scientific™ lab vision™
MS10340P1
500µL Each for $771.98

MS10340R7

 
thermo scientific™ lab vision™
MS10340R7
7mL Each for $647.16
Description & Specifications

Specifications

Antigen MUC-1
Clone SPM492
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Mouse
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Human breast cell line ZR-75-1
Isotype IgG2a kappa

Deliver accurate, reproducible results with the Thermo Scientific™ MUC-1, Mouse Monoclonal Antibody.

MUC-1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.

Host Species: Mouse

Clone: SPM492

Isotype: IgG1/k

Species Reactivity: Human. Others not tested.

Epitope: Not known

Immunogen: Human breast cell line ZR-75-1

Molecular Weight: 265-400kDa

Positive Control: Breast CA

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO