Learn More
Thiazolyl blue tetrazolium bromide, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 158990050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Thiazolyl blue tetrazolium bromide | |
298-93-1 | |
100.0 | |
97.5% min. (Argentometry) | |
C18H16BrN5S | |
5 g | |
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide | |
64965 | |
414.32 |
98% | |
97.5 | |
195.0°C | |
Glass bottle | |
MFCD00011964,MFCD00066662 | |
27,IV,6045 | |
Solubility in water: 10g/L (20°C). Other solubilities: soluble in 5% methanol, soluble in ethanol | |
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 | |
414.33 | |
CHEBI:53233 | |
98% |
Chemical Identifiers
298-93-1 | |
414.33 | |
AZKSAVLVSZKNRD-UHFFFAOYSA-M | |
64965 | |
[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1 |
C18H16BrN5S | |
MFCD00011964,MFCD00066662 | |
thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i | |
CHEBI:53233 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
GHS Signal Word: Warning
EINECSNumber : 206-069-5
RTECSNumber : XF8060000
TSCA : TSCA
RUO – Research Use Only