Please login to your online account to display your discounted pricing

MTPG, Tocris


Group II/group III mGlu antagonist; More selective for group II than group III

Quantity 50mg

Manufacturer: r & d systems  085550

 View more versions of this product

Catalog No. 085550

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 233.23
CAS 169209-66-9
Chemical Name or Material MTPG
Assay Percent Range >98%
Molecular Formula C10H11N5O2
Synonym (RS)-α-Methyl-4-tetrazolylphenylglycine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material MTPG
Assay Percent Range >98%
Molecular Formula C10H11N5O2
Formula Weight 233.23
Synonym (RS)-α-Methyl-4-tetrazolylphenylglycine
Accessories

Store at Room Temperature