missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.

$738.79 - $1805.97

Specifications

For Use With GC
Hazards Irritant
Storage 2°C to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Standard Components Price Quantity  
Catalog Number Mfr. No. Standard Components Price Quantity  
PI48920
View Documents Promotion Details
Thermo Scientific™
TS48920
MTBSTFA (N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide)
Pack for $738.79
 
PI48927
View Documents Promotion Details
Thermo Scientific™
TS48927
MTBSTFA +1% TBDMS (N-Methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide, Tertbutyldimetheylchlorosilane
Each for $1,805.97
 
Description

Description

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.

Additional Product Information:
  •  Derivatives have a high molecular concentration at M-57.
Specifications

Specifications

GC
Irritant
2°C to 8°C
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.