missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSTFA with 1% TMCS N-Methyl-N-(trimethylsilyl)trifluoroacetamide, ProteoSpec™ Ricca Chemical

GC Derivatization Reagent

Manufacturer:  Ricca Chemical Company RIRMB1204T10X1C2

Catalog No. RMB1204T101



Specifications

Specifications

ProteoSpec™ MSTFA with 1% TMCS N-Methyl-N-(trimethylsilyl)trifluoroacetamide
99,1
C6H12F3NOSi
CN(C(=O)C(F)(F)F)[Si](C)(C)C
199.248
CHEBI:85064
Liquid
Ampule
Colorless
24589-78-4,75-77-4
99,1
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
32510
199.25
Molecular Biology
1.11
10 x 1g

Chemical Identifiers

24589-78-4
199.248
32510
2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide
C6H12F3NOSi
MSPCIZMDDUQPGJ-UHFFFAOYSA-N
CHEBI:85064
CN(C(=O)C(F)(F)F)[Si](C)(C)C
Safety and Handling

Safety and Handling

ShelfLife : 12 months

Recommended Storage : Ambient Flammable

Documents