Please login to your online account to display your discounted pricing

MSPG, Tocris


Group II/group III mGlu antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS085410

 View more versions of this product

Catalog No. 085410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 245.25
Molecular Formula C9H11NO5S
CAS 169209-64-7
Chemical Name or Material MSPG
Assay Percent Range >98%
Synonym (RS)-α-Methyl-4-sulfonophenylglycine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material MSPG
Assay Percent Range >98%
Molecular Formula C9H11NO5S
Formula Weight 245.25
Synonym (RS)-α-Methyl-4-sulfonophenylglycine
Accessories

Store at Room Temperature