Please login to your online account to display your discounted pricing

MSPG, Tocris

Group II/group III mGlu antagonist

$238.81 - $984.19

Chemical Identifiers

Chemical Name or Material MSPG
Assay Percent Range >98%
Molecular Formula C9H11NO5S
Formula Weight 245.25
Synonym (RS)-α-Methyl-4-sulfonophenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

085410

 
r & d systems
RDS085410
10mg Each for $238.81

085450

 
r & d systems
RDS085450
50mg Each for $984.19
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 245.25
Chemical Name or Material MSPG
Assay Percent Range >98%
Molecular Formula C9H11NO5S
CAS 169209-64-7
Synonym (RS)-α-Methyl-4-sulfonophenylglycine
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature