Please login to your online account to display your discounted pricing

MSOP, Tocris


Specific group III mGlu antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS080350

 View more versions of this product

Catalog No. 080350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 199.1
CAS 66515-29-5
Synonym (RS)-α-Methylserine-O-phosphate
Chemical Name or Material MSOP
Assay Percent Range >98%
Molecular Formula C4H10NO6P
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material MSOP
Assay Percent Range >98%
Molecular Formula C4H10NO6P
Formula Weight 199.1
Synonym (RS)-α-Methylserine-O-phosphate
Accessories

Store at Room Temperature