Please login to your online account to display your discounted pricing

MSOP, Tocris

Specific group III mGlu antagonist

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material MSOP
Assay Percent Range >98%
Molecular Formula C4H10NO6P
Formula Weight 199.1
Synonym (RS)-α-Methylserine-O-phosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

080310

 
r & d systems
RDS080310
10mg Each for $244.81

080350

 
r & d systems
RDS080350
50mg Each for $997.23
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 199.1
Chemical Name or Material MSOP
Assay Percent Range >98%
Molecular Formula C4H10NO6P
CAS 66515-29-5
Synonym (RS)-α-Methylserine-O-phosphate
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature