Please login to your online account to display your discounted pricing
MSA™ Workman™ Construction Harnesses
Lightweight components and durable webbing for comfortable protection
$53.35 - $53.35
Specifications
Certifications/Compliance Meets ANSI A10.32 and Z359.1 standards, CSA Z259.10-M90 classes A and P, and all applicable OSHA standards including 1910.66 and 1926.502.
Color Yellow
Material Polyester Webbing
D-Ring Location Back
Size Standard
View More Specs
Products
Catalog Number Mfr. No. Buckle Style Price Quantity    
19-165-326
msa™
10072479
Qwik-Fit Each for $53.35
Description & Specifications
Specifications
Certifications/Compliance Meets ANSI A10.32 and Z359.1 standards, CSA Z259.10-M90 classes A and P, and all applicable OSHA standards including 1910.66 and 1926.502.
Color Yellow
Material Polyester Webbing
D-Ring Location Back
Size Standard
  • Abrasion-resistant hi-vis yellow polyester webbing
  • Zinc-plated, stamped alloy-steel D-rings and hardware
  • Retainer collars keep excess webbing neat
  • Integral back pad with removable tongue-buckle tool belt
  • Qwik-Fit™ chest strap on all models
  • Easy to don and adjust
Maximum Capacity: 400 lb. (181kg).
Meet ANSI™ A10.32 and Z359.1 standards, CSA Z259.10-M90 classes A and P, and all applicable OSHA standards including 1910.66 and 1926.502.