MSA™ V-Gard™ Slotted Protective Cap with Fas-Trac™ Suspension
Lightweight for all-day comfort
Manufacturer: MSA™ 475360
Alert:
Not specifically designed to protect against lateral blows from the front, side, or rear.
Description
- Polyethylene shell and suspension system work together as protection system
- Self-adjusting crown straps for comfortable fit
- Slotted design works with accessories
- Available in standard (61/2 to 8) size
- Four-point webbed Fas-Trac ratchet suspension with vinyl sweatband and large easy-adjustment knob
Order Info
Contact your Fisher Scientific Customer Service Representative for ordering information for sizes not shown.
Certifications
SEI certified.
Compliance
Meet or exceed all applicable requirements of ANSI™ Z89.1-2003 for Type I helmets, Class E and CSA Z94.1 2005.
Specifications
ANSI Z89.1-2014, CSA Z94.1-2015 | |
Polyethylene | |
Fas-Trac Ratchet |
Yellow | |
Standard |