missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MSA™ V-Gard™ Replacement Polycarbonate Visors

Help protect wearers from flying debris, splash or spray

$68.06 - $80.68

Specifications

Certifications/Compliance EN 166, ANSI, ISEA Z87.1-2010, AS, NZS 1337. 17-200-033, CSA Z94.3
Color Clear
For Use With (Application) Sanding and grinding, food processes, painting
Material Polycarbonate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Equipment) Product Type Size Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Product Type Size Price Quantity  
17-200-038 MSA™
10118094
V-Gard™ Frame Flat Visor 9.5 x 17 x 0.04 in. Case of 10 Each for $80.68
17-200-037 MSA™
10117783
V-Gard™ Retractable or Standard Chin Protector Flat Visor 8 x 17 x 0.06 in. Case of 10 Each for $68.06
Specifications

Description

For use with V-Gard frames only; fit on all MSA Helmets.

  • Clear polycarbonate offers maximum light transmission and can be used indoors, outdoors and in low-light conditions
  • Maximum UV protection
  • Nitrometer style can be used only with V-Gard retractable or standard chin protector

Compliance

All visors meet EN 166, ANSI/ISEA Z87.1-2010 and AS/NZS 1337. 17-200-033 also meets CSA Z94.3.

Description & Specifications

Specifications

EN 166, ANSI, ISEA Z87.1-2010, AS, NZS 1337. 17-200-033, CSA Z94.3
Clear
Sanding and grinding, food processes, painting
Polycarbonate