MSA™ V-Gard™ Protective Slotted Cap with Staz-On™ Suspension
Lightweight for all-day wearing comfort
Manufacturer:  MSA™ 475235
 View more versions of this product
Catalog No. 2985321
Description
Description
  • Four-point webbed Staz-On suspension with perforated vinyl sweatband
  • Sizes: Small (6-7 1/8), Standard (6 1/2-8), and Large (7-8 1/2)
  • Available in more than 20 colors, with custom imprinting and striping available
Order Info
Contact your Fisher Safety Customer Safety Representative for ordering information for sizes and colors not shown.
†With lamp bracket and cord holder (Class C)
Specifications
Specifications
Safety
Black
Staz-On Pin-Lock
Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
Standard
Documents