MSA™ V-Gard™ Slotted Full-Brim Hats
Full-brim hardhat available in multiple, fade-resistant colors
Manufacturer: MSA™ 475366
Description
- Polyethylene shell provides superior impact protection
- Self-adjusting crown straps ensure a comfortable fit
- Lightweight and well-balanced for all-day wearing comfort
- Slotted design works with MSA accessories
- Standard size (6½ to 8)
Suspension System Options:
- 1-Touch™ Suspension
- One-handed squeeze opening
- One-hand slide adjustment
- Snap closure
- Adjustable dual nylon straps
- Staz-On Suspension
- 4-point suspension system helps keep helmet in place
- Features high-quality sweatband and nylon height adjusters
- Easily clips into V Gard hat or cap shell
- Adjustable liner
- Fas-Trac III Suspension
- Flush rear lug attachments eliminate pressure points
- Three levels of nape strap adjustment for customized fit
- Graspable ratchet with ultra-smooth rotation and steadfast hold
Specifications
Yellow | |
Standard | |
Fas-Trac ratchet suspension |